4,4'-(Buta-1,3-diyne-1,4-diyl)dianiline 97%

Supplier: Ambeed
Warning

A715767-100MG A715767-1G A715767-250MG A715767-5G
77198-946EA 33.99 USD
77198-946 77198-948 77198-950 77198-952
4,4'-(Buta-1,3-diyne-1,4-diyl)dianiline 97%
4,4'-(Buta-1,3-diyne-1,4-diyl)dianiline

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.27 g/mol
CAS Number: 30405-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR