3-Methyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A715126-1G A715126-250MG
77198-938EA 326.99 USD
77198-938 77198-940
3-Methyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 97%
3-Methyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃
MW: 137.18 g/mol
MDL Number: MFCD16619563
CAS Number: 126052-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR