3-(Hydroxymethyl)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Benzenedimethanol, m-Xylene-α,α'-diol, m-Phenylenedimethanol

A714631-100G A714631-10G A714631-1G A714631-5G
77198-930EA 248.23 USD
77198-930 77198-932 77198-934 77198-936
3-(Hydroxymethyl)benzyl alcohol 98%
3-(Hydroxymethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00004649
CAS Number: 626-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR