1-Benzyl-5-nitro-1H-imidazole 95%

Supplier: Ambeed
Warning

A714046-1G A714046-5G
77198-926EA 46 USD
77198-926 77198-928
1-Benzyl-5-nitro-1H-imidazole 95%
1-Benzyl-5-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O₂
MW: 203.20 g/mol
MDL Number: MFCD04972447
CAS Number: 159790-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR