1,4-Pentanediol 98%

Supplier: Ambeed
Warning

A713889-1G A713889-25G A713889-500G A713889-5G
77198-918EA 12.36 USD
77198-918 77198-920 77198-922 77198-924
1,4-Pentanediol 98%
1,4-Pentanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MW: 104.149 g/mol
MDL Number: MFCD00004560
CAS Number: 626-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR