2-(4-Bromophenyl)propan-2-amine hydrochloride 98%

Supplier: Ambeed
Warning

A712615-1G A712615-250MG
77170-806EA 92.7 USD
77170-806 77170-808
2-(4-Bromophenyl)propan-2-amine hydrochloride 98%
2-(4-Bromophenyl)propan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃BrClN
MW: 250.56 g/mol
MDL Number: MFCD11505546
CAS Number: 1173047-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR