tert-Butyl-3-hydroxy-3-methylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-hydroxy-3-methylpiperidine-1-carboxylate

A710989-100MG A710989-1G A710989-250MG A710989-5G
77170-792EA 16.48 USD
77170-792 77170-794 77170-796 77684-628
tert-Butyl-3-hydroxy-3-methylpiperidine-1-carboxylate 95%
tert-Butyl-3-hydroxy-3-methylpiperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD13194152
CAS Number: 1104083-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR