SB242235 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(4-Fluorophenyl)-1-(piperidin-4-yl)-1H-imidazol-5-yl)-2-methoxypyrimidine, SB 242235, 4-[5-(4-fluorophenyl)-3-piperidin-4-ylimidazol-4-yl]-2-methoxypyrimidine

A710495-10MG A710495-1MG A710495-5MG
77170-780EA 103 USD
77170-780 77170-782 77170-784
SB242235 98%
SB242235

Precaución: for Research Use Only

Formula: C₁₉H₂₀FN₅O
MW: 353.4 g/mol
MDL Number: MFCD08690575
CAS Number: 193746-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR