(S)-(+)-1-Fmoc-3-pyrrolidinol 95%

Supplier: Ambeed
Warning

Synonyms: (S)-1-fmoc-3-pyrrolidinol

A709157-100MG A709157-1G A709157-250MG A709157-5G
77198-894EA 24 USD
77198-894 77198-896 77198-898 77198-900
(S)-(+)-1-Fmoc-3-pyrrolidinol 95%
(S)-(+)-1-Fmoc-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₃
MDL Number: MFCD01317842
CAS Number: 215178-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR