1,2-Hexadecanediol 98%

Supplier: Ambeed
Warning

A709054-1G A709054-25G A709054-5G
77198-888EA 8 USD
77198-888 77198-890 77198-892
1,2-Hexadecanediol 98%
1,2-Hexadecanediol

Precaución: for Research Use Only

Formula: C₁₆H₃₄O₂
MDL Number: MFCD00004727
CAS Number: 6920-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR