α-Methyl-4-trifluoromethylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethanol

A708197-100G A708197-1G A708197-25G A708197-5G
77198-880EA 211.01 USD
77198-880 77198-882 77198-884 77198-886
α-Methyl-4-trifluoromethylbenzyl alcohol 97%
alpha-Methyl-4-trifluoromethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MDL Number: MFCD00039618
CAS Number: 1737-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR