5-Bromo-1-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A708140-1G A708140-250MG
77170-774EA 126.69 USD
77170-774 77170-776
5-Bromo-1-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-1-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₉BrN₂
MW: 213.07 g/mol
MDL Number: MFCD22394541
CAS Number: 1187421-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR