1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-tert-Butyl 2-methyl 4-bromo-1H-pyrrole-1,2-dicarboxylate
A707947-100MG
A707947-250MG
77198-876EA
36.05
USD
77198-876
77198-878
1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate 97%
1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄BrNO₄ MW: 304.15 g/mol |
MDL Number:
MFCD09751125 CAS Number: 156237-78-4 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...