2-(2,3-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%

Supplier: Ambeed
Warning

A705716-100MG A705716-1G A705716-250MG A705716-5G
77170-756EA 6 USD
77170-756 77170-758 77170-760 77569-344
2-(2,3-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%
2-(2,3-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₂H₁₅BCl₂O₂
MDL Number: MFCD18729912
CAS Number: 1255760-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR