4-Acetamidobutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Acetamidobutanoic acid

A705709-1G A705709-5G A705709-25G
77170-752EA 16.48 USD
77170-752 77170-754 77684-618
4-Acetamidobutyric acid 98%
4-Acetamidobutyric acid

Precaución: For research use only.

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
MDL Number: MFCD00020552
CAS Number: 3025-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR