6-Bromo-5-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A705701-1G A705701-250MG
77198-858EA 253.38 USD
77198-858 77198-860
6-Bromo-5-nitro-1H-indazole 95%
6-Bromo-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
MDL Number: MFCD27987808
CAS Number: 1351813-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR