7,7-Dimethyl-2,3,4,6,7,8-hexahydro-1H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-1-one 95%

Supplier: Ambeed
Warning

A704497-10G A704497-1G A704497-250MG A704497-5G
77198-852EA 879.62 USD
77198-852 77198-854 77198-856 77747-408
7,7-Dimethyl-2,3,4,6,7,8-hexahydro-1H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-1-one 95%
7,7-Dimethyl-2,3,4,6,7,8-hexahydro-1H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-1-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
MDL Number: MFCD24619432
CAS Number: 1346674-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR