1-(4-Fluorophenyl)cyclopropylamine 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Fluorophenyl)cyclopropanamine
A704405-100MG
A704405-250MG
A704405-50MG
A704405-1G
A704405-5G
77170-746EA
25
USD
77170-746
77170-748
77170-750
77684-615
77684-616
1-(4-Fluorophenyl)cyclopropylamine 98%, Ambeed.Inc
1-(4-Fluorophenyl)cyclopropylamine
Precaución: For research use only.
Formula:
C₉H₁₀FN MW: 151.181 g/mol |
MDL Number:
MFCD07374474 CAS Number: 474709-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...