tert-Butyl 4-(6-bromopyridin-3-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A704211-250MG A704211-5G A704211-100MG A704211-1G
77170-742EA 28 USD
77170-742 77170-744 77893-414 77893-415
tert-Butyl 4-(6-bromopyridin-3-yl)piperazine-1-carboxylate 95%
tert-Butyl 4-(6-bromopyridin-3-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrN₃O₂
MDL Number: MFCD11872605
CAS Number: 412348-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR