Ethyl 2-(5-chloropyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A702468-100MG A702468-250MG
77170-726EA 74.16 USD
77170-726 77614-174
Ethyl 2-(5-chloropyridin-2-yl)acetate 95%
Ethyl 2-(5-chloropyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD10698594
CAS Number: 1060814-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR