tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
Supplier: Ambeed
|
Danger
|
A702139-10G
A702139-1G
A702139-250MG
77170-720EA
962.02
USD
77170-720
77170-722
77170-724
tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₆BrN₃O₂ MW: 302.17 g/mol |
MDL Number:
MFCD17480368 CAS Number: 877399-34-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...