1-(2-Cyanoethyl)-2-methylimidazole 96%

Supplier: Ambeed
Danger

A702120-1G A702120-250MG A702120-25G A702120-5G
77198-844EA 14 USD
77198-844 77198-846 77198-848 77198-850
1-(2-Cyanoethyl)-2-methylimidazole 96%
1-(2-Cyanoethyl)-2-methylimidazole

Precaución: for Research Use Only

Formula: C₇H₉N₃
MDL Number: MFCD00047049
CAS Number: 23996-55-6
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR