Methyl 2-(2-aminobenzo[d]thiazol-6-yl)acetate 97%
Supplier: Ambeed
|
Warning
|
A701819-100MG
A701819-1G
A701819-5G
77198-842EA
40.17
USD
77198-842
77569-338
77573-268
Methyl 2-(2-aminobenzo[d]thiazol-6-yl)acetate 97%
Methyl 2-(2-aminobenzo[d]thiazol-6-yl)acetate
Precaución: For research use only.
Formula:
C₁₀H₁₀N₂O₂S MW: 222.26 g/mol |
MDL Number:
MFCD18643368 CAS Number: 99738-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...