Pentafluorophenoxyacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Perfluorophenoxy)acetic acid
A700526-1G
A700526-25G
A700526-5G
77198-826EA
19
USD
77198-826
77198-828
77198-830
Pentafluorophenoxyacetic acid 98%
Pentafluorophenoxyacetic acid
Precaución: for Research Use Only
Formula:
C₈H₃F₅O₃ MW: 242.10 g/mol |
MDL Number:
MFCD00004297 CAS Number: 14892-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...