5-(2-Pyridyl)-1H-tetrazole 98%

Supplier: Ambeed
Danger

Synonyms: 2-(1H-Tetrazol-5-yl)pyridine, 5-(2-Pyridyl)-1H-tetrazole

A699027-1G A699027-250MG A699027-5G
77170-708EA 23.69 USD
77170-708 77170-710 77552-118
5-(2-Pyridyl)-1H-tetrazole 98%
5-(2-Pyridyl)-1H-tetrazole

Precaución: for Research Use Only

Formula: C₆H₅N₅
MW: 147.14 g/mol
MDL Number: MFCD00068114
CAS Number: 33893-89-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR