N4-Cyclopropyl-6-(2,3-dichlorophenyl)pyrimidine-2,4-diamine 98%

Supplier: Ambeed
Warning

Synonyms: TH588

A698512-1MG A698512-25MG A698512-5MG
77170-702EA 12.36 USD
77170-702 77170-704 77170-706
N4-Cyclopropyl-6-(2,3-dichlorophenyl)pyrimidine-2,4-diamine 98%
N4-Cyclopropyl-6-(2,3-dichlorophenyl)pyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₁₃H₁₂Cl₂N₄
MW: 295.17 g/mol
MDL Number: MFCD28963908
CAS Number: 1609960-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR