(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) 95%

Supplier: Ambeed
Warning

Synonyms: N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)-1-naphthamide), (R,R)-DACH-Naphthyl Trost Ligand

A698261-100MG A698261-1G A698261-250MG A698261-5G
77170-696EA 30.9 USD
77170-696 77170-698 77170-700 77684-610
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) 95%
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl)

Precaución: For research use only.

Formula: C₅₂H₄₄N₂O₂P₂
MW: 790.8655 g/mol
MDL Number: MFCD02684551
CAS Number: 174810-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR