tert-Butyl (6-chloropyridazin-3-yl)carbamate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A698226-1G A698226-5G A698226-25G
77170-694EA 35 USD
77170-694 77614-158 77684-609
tert-Butyl (6-chloropyridazin-3-yl)carbamate 95%, Ambeed.Inc
tert-Butyl (6-chloropyridazin-3-yl)carbamate

Precaución: For research use only.

Formula: C₉H₁₂ClN₃O₂
MW: 229.66 g/mol
MDL Number: MFCD18070957
CAS Number: 1276056-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR