Methyl 3-chloro-5-methylbenzoate 95%
Supplier: Ambeed
|
Warning
|
A698159-5G
A698159-1G
77170-692EA
168.92
USD
77170-692
77684-608
Methyl 3-chloro-5-methylbenzoate 95%
Methyl 3-chloro-5-methylbenzoate
Precaución: For research use only.
Formula:
C₉H₉ClO₂ MW: 184.61 g/mol |
MDL Number:
MFCD11007828 CAS Number: 153203-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...