6-Chloro-2-fluoropyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A698098-100MG A698098-1G A698098-250MG A698098-5G
77170-684EA 17 USD
77170-684 77170-686 77170-688 77170-690
6-Chloro-2-fluoropyridine-3-carbaldehyde 97%
6-Chloro-2-fluoropyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MDL Number: MFCD16556251
CAS Number: 1093880-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR