1,4-Butanedithiol 99+%

Supplier: Ambeed
Danger

Synonyms: 1,4-Dimercaptobutane

A698021-25G A698021-5G
77198-810EA 65 USD
77198-810 77198-812
1,4-Butanedithiol 99+%
1,4-Butanedithiol

Precaución: for Research Use Only

Formula: C₄H₁₀S₂
MW: 122.26 g/mol
Boiling Pt: 105
Melting Pt: -53
MDL Number: MFCD00004906
CAS Number: 1191-08-8
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR