1-(2,3-Dihydro-1H-indol-5-yl)ethanone 95%
Supplier: Ambeed
|
Warning
|
A697829-250MG
A697829-1G
77198-808EA
17
USD
77198-808
77569-328
1-(2,3-Dihydro-1H-indol-5-yl)ethanone 95%
1-(2,3-Dihydro-1H-indol-5-yl)ethanone
Precaución: For research use only.
Formula:
C₁₀H₁₁NO |
MDL Number:
MFCD00090836 CAS Number: 16078-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...