(4-Bromo-2-isopropylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-isopropylphenylboronic acid

A696934-100MG A696934-1G A696934-250MG
77198-788EA 56 USD
77198-788 77198-790 77198-792
(4-Bromo-2-isopropylphenyl)boronic acid 98%
(4-Bromo-2-isopropylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BBrO₂
MDL Number: MFCD22628234
CAS Number: 1451390-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR