5-Fluoro-7-azaindole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

A696862-1G A696862-250MG
77170-668EA 54 USD
77170-668 77170-670
5-Fluoro-7-azaindole-3-carboxylic acid 97%
5-Fluoro-7-azaindole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MDL Number: MFCD12962623
CAS Number: 1067193-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR