2-Phenyl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Warning
|
A695208-100MG
A695208-1G
A695208-250MG
A695208-5G
77170-658EA
39
USD
77170-658
77170-660
77170-662
77893-389
2-Phenyl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
2-Phenyl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂BF₄N₃ |
MDL Number:
MFCD29059016 CAS Number: 909122-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...