2,3-Dimethylphenylhydrazinium hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,3-Dimethylphenyl)hydrazine hydrochloride, 2,3-Dimethylphenylhydrazine hydrochloride, (2,3-Dimethylphenyl)hydrazine hydrochloride

A694752-1G A694752-5G
77198-778EA 7 USD
77198-778 77198-780
2,3-Dimethylphenylhydrazinium hydrochloride 95%
2,3-Dimethylphenylhydrazinium hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MDL Number: MFCD00013380
CAS Number: 56737-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR