2-(Aminosulfonyl)-N,N-dimethylnicotinamide 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-2-sulphamoylnicotinamide

A694435-1G A694435-250MG A694435-5G
77198-772EA 12.36 USD
77198-772 77198-774 77198-776
2-(Aminosulfonyl)-N,N-dimethylnicotinamide 95%
2-(Aminosulfonyl)-N,N-dimethylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O₃S
MW: 229.26 g/mol
MDL Number: MFCD03265507
CAS Number: 112006-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR