2,2'-Diamino-4,4'-bithiazole 98%

Supplier: Ambeed
Warning

Synonyms: [4,4'-Bithiazole]-2,2'-diamine, 2,2'-Diamino-4,4'-bis(1,3-thiazole), 4,4'-Bithiazole-2,2'-diamine

A693142-100MG A693142-1G A693142-250MG A693142-5G
77198-752EA 49.44 USD
77198-752 77198-754 77198-756 77198-758
2,2'-Diamino-4,4'-bithiazole 98%
2,2'-Diamino-4,4'-bithiazole

Precaución: for Research Use Only

Formula: C₆H₆N₄S₂
MW: 198.26 g/mol
MDL Number: MFCD01740587
CAS Number: 58139-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR