Morpholino(1H-pyrazol-4-yl)methanone 97%

Supplier: Ambeed
Warning

A692724-1G A692724-250MG
77198-748EA 396.55 USD
77198-748 77198-750
Morpholino(1H-pyrazol-4-yl)methanone 97%
Morpholino(1H-pyrazol-4-yl)methanone

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O₂
MW: 181.19 g/mol
MDL Number: MFCD12142788
CAS Number: 1152749-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR