3-Fluoro-α-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

A692251-1G A692251-25G A692251-5G
77198-732EA 24.72 USD
77198-732 77198-734 77198-736
3-Fluoro-α-methylbenzyl alcohol 97%
3-Fluoro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD00014402
CAS Number: 402-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR