1-(2-Thienyl)-1-propanone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Propionylthiophene

A691949-25G A691949-5G
77198-728EA 27.81 USD
77198-728 77198-730
1-(2-Thienyl)-1-propanone 98%
1-(2-Thienyl)-1-propanone

Precaución: for Research Use Only

Formula: C₇H₈OS
MW: 140.20 g/mol
MDL Number: MFCD00005446
CAS Number: 13679-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR