2-Acetyl-4-bromothiophene 97%
Supplier: Ambeed
|
Warning
|
A691829-100G
A691829-10G
A691829-1G
A691829-250MG
A691829-25G
A691829-5G
77198-716EA
339.9
USD
77198-716
77198-718
77198-720
77198-722
77198-724
77198-726
2-Acetyl-4-bromothiophene 97%
2-Acetyl-4-bromothiophene
Precaución: for Research Use Only
Formula:
C₆H₅BrOS MW: 205.08 g/mol |
MDL Number:
MFCD01566552 CAS Number: 7209-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...