4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dihydroxy-5-methylisophthalaldehyde

A690828-1G A690828-250MG
77198-712EA 69.01 USD
77198-712 77198-714
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 97%
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.157 g/mol
MDL Number: MFCD11505923
CAS Number: 22304-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR