(S)-5-Methylpiperazin-2-one 98%

Supplier: Ambeed
Danger

A690093-250MG A690093-100MG
77198-710EA 189 USD
77198-710 77747-388
(S)-5-Methylpiperazin-2-one 98%
(S)-5-Methylpiperazin-2-one

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O
MDL Number: MFCD07373484
CAS Number: 1240583-20-3
UN: 3263
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR