tert-Butyl methyl(6-methylpyridin-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A689226-1G A689226-250MG A689226-5G
77198-704EA 35 USD
77198-704 77198-706 77198-708
tert-Butyl methyl(6-methylpyridin-2-yl)carbamate 95%
tert-Butyl methyl(6-methylpyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD08275048
CAS Number: 205676-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR