Isopropyl 2-hydroxy-2-phenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Pregabalin impurity D

A687252-1G A687252-5G A687252-250MG
77170-618EA 472.01 USD
77170-618 77893-376 77893-375
Isopropyl 2-hydroxy-2-phenylacetate 98%
Isopropyl 2-hydroxy-2-phenylacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MDL Number: MFCD01715837
CAS Number: 4118-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR