trans-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A687098-100MG
A687098-250MG
A687098-1G
77170-608EA
180.25
USD
77170-608
77170-610
77684-596
trans-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid 98%
trans-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid
Precaución: For research use only.
Formula:
C₁₁H₁₈FNO₄ MW: 247.26 g/mol |
MDL Number:
MFCD28016233 CAS Number: 1903422-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...