4-Fluoro-2-nitrobenzyl alcohol 98+%

Supplier: Ambeed
Warning

A686499-250MG A686499-1G
77170-596EA 10 USD
77170-596 77893-373
4-Fluoro-2-nitrobenzyl alcohol 98+%
4-Fluoro-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD07368344
CAS Number: 1043416-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR