1,2-Bis(3-indenyl)ethane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,3'-Ethylenebis(1H-indene), 1,2-Di(1H-inden-3-yl)ethane
A686336-1G
A686336-25G
A686336-5G
77198-696EA
13
USD
77198-696
77198-698
77198-700
1,2-Bis(3-indenyl)ethane 98%
1,2-Bis(3-indenyl)ethane
Precaución: for Research Use Only
Formula:
C₂₀H₁₈ MW: 258.36 g/mol |
MDL Number:
MFCD00191814 CAS Number: 18657-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...