Octane-1,8-diamine, N-BOC protected 97%

Supplier: Ambeed
Warning

A683766-100MG A683766-1G A683766-250MG A683766-5G
77198-686EA 10 USD
77198-686 77198-688 77198-690 77198-692
Octane-1,8-diamine, N-BOC protected 97%
Octane-1,8-diamine, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₃H₂₈N₂O₂
MDL Number: MFCD02094499
CAS Number: 88829-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR